Learn More
Biosan Laboratories Sani-Check YM Kit, 25 tests
Supplier:  Biosan Laboratories 120
Encompass_Preferred
The growth of fungi (yeast and mold) in the industrial environment is often responsible for extensive losses in productivity. Problems related to high levels of mold include slime formation, offensive odors and plugging of fluid delivery lines. Effective control of fungi through biocide use is only possible when routine monitoring of fungal levels is performed. Sani-Check YM Test Kit for yeast and mold makes routine determinations of fungal populations quick and easy.
Catalog No. NC0523146