Learn More
Cole-Parmer Global Water WS700 Peristaltic Water Sampler
Supplier: Cole-Parmer 9924370
Simple to operate; no programming required. Automatic 15-second backflush clears pickup strainer and hose. Optional kit available for storm water sampling.