Learn More
Hach Company Sampler, Water Core, "Sludge Judge" Large Capacity, 1-1/4" Diameter
Supplier:  Hach Company 2506102
Encompass_Preferred
Accurate readings can be obtained on suspended solids up to a depth of 4.5 m (15'). Comes in three 1.5 m (5') sections with screw-type fittings. Designed for sampling solids in wastewater and food processing applications. Standard Sludge Judge features 3/4" diameter plastic tubing, while the large capacity Sludge Judge features 1-1/4" diameter plastic tubing for double the capacity.
Catalog No. 50-199-6107