Learn More
Electron Microscopy Sciences K-kits Sample Loading Stage
Supplier: Electron Microscopy Sciences K7264
Loading Stage consists of an anodized aluminum body. It has a horizontal bar in a recess on the side and a hole in the middle to house the Liquid Stage, which is a removable stainless steel rod. For use with K-kit TEM observation of liquid samples.