Learn More
Alpco Ltd Salivette Collection Devices
Supplier: Alpco Ltd 12BSVC50U
The Salivettes (Saliva Collection Devices) contain a sterile cotton swab, label, and exterior tube for collection of saliva samples prior to assay. For simple, hygienic collection and storage of saliva samples. Each box contains 50 units. Research Use Only. Not for Use in Diagnostic Procedures.
This item is not returnable.
View return policy