Learn More
Zymo Research Corporation DNA/RNA Shield SafeCollect Saliva Collection Kit - CE
SDP
Supplier:  Zymo Research Corporation R1211E
Encompass_Preferred
User-friendly saliva collection for stabilizing the nucleic acid content of saliva and sputum samples. DNA/RNA Shield completely inactivates harmful pathogens in the sample, and stabilizes the RNA in a sample for up to 30 days at room temperature. The SafeCollect design eliminates the risk of spilling the stabilization reagent during use, and is ideal for applications that require at-home self collection.
Catalog No. NC2021757