Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hanna Pharmaceutical Supply Co., Inc. 0.9% SODIUM CHLOR. INJ.10ML

Supplier:  Hanna Pharmaceutical Supply Co., Inc. 0409488810

Encompass_Preferred

Sodium Chloride 0.9% Inj.USP 10ml ftv (flip top vial) 25/PK
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.

Catalog No. NC9054335


Add to Cart
This item is not returnable. View return policy

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit