Learn More
Bioassay Systems QuantiChrom™ Salicylate Assay Kit. For quantitative determination of salicylate in biological samples and beauty products, mouthwash, etc.
SDP
Supplier:  Bioassay Systems DSALY100
Encompass_Preferred
For quantitative determination of salicylate in biological samples and beauty products, mouthwash, etc. Key Features: Fast and sensitive. Linear detection range: 0.8 mM (10.9 mg/dL) to 20 mM (274.2 mg/dL) salicylate with 20 µL sample (96-well). Convenient. The procedure involves adding a single working reagent. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well or 384-well plate assay for thousands of samples per day. Method: OD560nm. Samples: Serum, plasma, urine, food, beverage, agriculture, drugs, beauty, mouthwash samples, etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 Tests.
Catalog No. 50-225-5866