Learn More
Accuform™ Safety Signs
Accuform™ Safety Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
$11.73 - $17.09
Specifications
Certifications/Compliance | OSHA 29, CFR 1910.145, and ANSI Z535 |
---|---|
Material | Plastic |
Mounting Type | Corner Holes |
Color | Black on Yellow |
Catalog Number | Mfr. No. | Displayed Text | dimLanguages | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Displayed Text | dimLanguages | Price | Quantity & Availability | ||||
19-169-579
|
Accuform™
MCHL645VP |
CAUTION HAZARDOUS WASTE SATELLITE ACCUMULATION POINT |
Each for $11.73
|
|
|||||
19-169-580
|
Accuform™
MCHL594VP |
SATELLITE STORAGE AREA |
Each for $17.09
|
|
|||||
Description
- Durable plastic has slight flex that is ideal for indoor and sheltered outdoor environments
- Printed surface has clear UV protective overcoat
- 50% recycled content with UV inhibitors and good impact resistance
- Rounded corners with 3/16 in. mounting holes for installation
- Comply in accordance with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
Specifications
OSHA 29, CFR 1910.145, and ANSI Z535 | |
Plastic | |
Corner Holes | |
Black on Yellow |