Learn More
Radiometer America Radiometer Rinse Solution for ABL800 FLEX Blood Gas Analyzer
Supplier: Radiometer America 944130
ABL800 FLEX Rinse Solution (S4980)
Rinse Solution for the ABL800 FLEX blood gas analyzer.
ABL800 FLEX Blood Gas Analyzer
With 15,000 active analyzers globally, the ABL800 FLEX analyzer provides you with proven performance. The accuracy and precision of the analyzer are based on advanced sensor technology, an optical measuring system and the fact that several of the steps normally associated with blood gas testing are fully automated when using the ABL800 FLEX analyzer. As a result, you get high analytical performance in the measurement of pH, blood gases, electrolytes, metabolites and oximetry parameters.
This item has a minimum order quantity of 2 per supplier requirements.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.