Learn More
Horiba Instruments S010 REPLACEMENT PH SENSOR
Supplier: Horiba Instruments 3200459834
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Replacement sensor for the B-713, PH11, PH22, PH33 Employing the same test principle as standard laboratory electrodes, the LAQUAtwin packs all components into a flat sensor that’s less than 1 mm thick. Waterproof construction protects the instrument.