Learn More
Labconco™ RXPert™ Double Filtered Balance Systems
![SDP](/content/dam/fishersci/glyphs/sdp.png)
The patented RXPert Filtered Balance System is Class I Enclosure that meets USP <800> requirements for non-sterile compounding when ducted. Models feature 2 HEPA filters to provide redundant HEPA filtration in series and therefore require NO ductwork.
$14810.00 - $36100.00
Specifications
Product Type | Balance Enclosure |
---|---|
Lighting Type | Fluorescent |
Certifications/Compliance | Certifications: ANSI Z9.5, CAN/CSA C22.2, Modified ASHRAE 110, SEFA 1, UL 61010 |
Hertz | 60 Hz |
Voltage | 115 V |
Catalog Number | Mfr. No. | Dimensions (D x W x H) | Includes | Weight (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (D x W x H) | Includes | Weight (English) | Price | Quantity & Availability | |||
10-305-023
![]() |
Labconco™
3971280 |
23.4 x 21.6 x 22.7 in. (59.4 x 54.8 x 57.6 cm) | – | 269 lb. |
Each for $14,810.00
|
|
|||
10-305-024
![]() |
Labconco™
3971281 |
23.4 x 21.6 x 22.7 in. (59.4 x 54.8 x 57.6 cm) | 344 lb. |
Each for $16,170.00
|
|
||||
10-305-025
![]() |
Labconco™
3971282 |
23.4 x 21.6 x 22.7 in. (59.4 x 54.8 x 57.6 cm) | 364 lb. |
Each for $15,930.00
|
|
||||
10-305-026
![]() |
Labconco™
3971380 |
23.4 x 33.6 x 22.7 in. (59.4 x 85.3 x 57.6 cm) | – | 327 lb. |
Each for $17,230.00
|
|
|||
10-305-027
![]() |
Labconco™
3971381 |
23.4 x 33.6 x 22.7 in. (59.4 x 85.3 x 57.6 cm) | 401 lb. |
Each for $18,500.00
|
|
||||
10-305-028
![]() |
Labconco™
3971382 |
23.4 x 33.6 x 22.7 in. (59.4 x 85.3 x 57.6 cm) | Epoxy resin work surface | 452 lb. |
Each for $17,290.00
|
|
|||
10-305-029
![]() |
Labconco™
3971480 |
23.4 x 45.6 x 22.7 in. (59.4 x 115.8 x 57.6 cm) | – | 104 lb. |
Each for $16,730.00
|
|
|||
10-305-030
![]() |
Labconco™
3971481 |
23.4 x 45.6 x 22.7 in. (59.4 x 115.8 x 57.6 cm) | 501 lb. |
Each for $20,060.00
|
|
||||
10-305-031
![]() |
Labconco™
3971482 |
23.4 x 45.6 x 22.7 in. (59.4 x 115.8 x 57.6 cm) | 541 lb. |
Each for $19,840.00
|
|
||||
10-305-032
![]() |
Labconco™
3971485 |
23.4 x 45.6 x 32 in. (59.4 x 115.8 x 81.2 cm) | – | 420 lb. |
Each for $18,380.00
|
|
|||
10-305-033
![]() |
Labconco™
3971486 |
23.4 x 45.6 x 32 in. (59.4 x 115.8 x 81.2 cm) | 520 lb. |
Each for $20,850.00
|
|
||||
10-305-034
![]() |
Labconco™
3971487 |
23.4 x 45.6 x 32 in. (59.4 x 115.8 x 81.2 cm) | 560 lb. |
Each for $20,720.00
|
|
||||
10-305-035
![]() |
Labconco™
3971490 |
30 x 45.6 x 32 in. (76.2 x 115.8 x 81.2 cm) | – | 450 lb. |
Each for $18,380.00
|
|
|||
10-305-036
![]() |
Labconco™
3971491 |
30 x 45.6 x 32 in. (76.2 x 115.8 x 81.2 cm) | 550 lb. |
Each for $20,850.00
|
|
||||
10-305-037
![]() |
Labconco™
3971492 |
30 x 45.6 x 32 in. (76.2 x 115.8 x 81.2 cm) | 590 lb. |
Each for $19,830.00
|
|
||||
10-305-038
![]() |
Labconco™
3971585 |
23.4 x 57.6 x 32 in. (59.4 x 146.3 x 81.2 cm) | – | 698 lb. |
Each for $24,990.00
|
|
|||
10-305-039
![]() |
Labconco™
3971586 |
23.4 x 57.6 x 32 in. (59.4 x 146.3 x 81.2 cm) | 798 lb. |
Each for $27,900.00
|
|
||||
10-305-040
![]() |
Labconco™
3971587 |
23.4 x 57.6 x 32 in. (59.4 x 146.3 x 81.2 cm) | 838 lb. |
Each for $27,200.00
|
|
||||
10-305-041
![]() |
Labconco™
3971590 |
30 x 57.6 x 32 in. (76.2 x 146.3 x 81.2 cm) | – | 745 lb. |
Each for $28,200.00
|
|
|||
10-305-042
![]() |
Labconco™
3971591 |
30 x 57.6 x 32 in. (76.2 x 146.3 x 81.2 cm) | 845 lb. |
Each for $31,150.00
|
|
||||
10-305-043
![]() |
Labconco™
3971592 |
30 x 57.6 x 32 in. (76.2 x 146.3 x 81.2 cm) | 885 lb. |
Each for $31,900.00
|
|
||||
10-305-044
![]() |
Labconco™
3971685 |
23.4 x 69.6 x 32 in. (59.4 x 176.7 x 81.2 cm) | – | 840 lb. |
Each for $27,200.00
|
|
|||
10-305-045
![]() |
Labconco™
3971686 |
23.4 x 69.6 x 32 in. (59.4 x 176.7 x 81.2 cm) | 940 lb. |
Each for $32,650.00
|
|
||||
10-305-046
![]() |
Labconco™
3971687 |
23.4 x 69.6 x 32 in. (59.4 x 176.7 x 81.2 cm) | 890 lb. |
Each for $30,300.00
|
|
||||
10-305-047
![]() |
Labconco™
3971690 |
30 x 69.6 x 32 in. (76.2 x 176.7 x 81.2 cm) | – | 892 lb. |
Each for $30,450.00
|
|
|||
10-305-048
![]() |
Labconco™
3971691 |
30 x 69.6 x 32 in. (76.2 x 176.7 x 81.2 cm) | 992 lb. |
Each for $33,700.00
|
|
||||
10-305-049
![]() |
Labconco™
3971692 |
30 x 69.6 x 32 in. (76.2 x 176.7 x 81.2 cm) | 1032 lb. |
Each for $36,100.00
|
|
||||
Description
- Quiet operation
- HEPA Filtered Enclosure for powder weighing
- Two redundant filters
- Each HEPA filter 100% independently leak-detection scannable without removing impellers or filters
- Bag-in/bag-out HEPA filter for protection during filter changes
- Glass and steel construction
- Ergonomic air foil with aerodynamic patented Clean-Sweep airflow openings
- Guardian airflow monitor that continuously monitors airflow with visable and audible alarms when airflow is outside set point range
- Upper dilution air supply and upper containment sash foil
- Zone-perforated, pivoting rear baffle
- Side-entry air foils
- Intrinsically safe, negative pressure design
- Built-in blower with dynamically-balanced, vibration-isolated motorized impeller and speed control
- Blower has a 40% reserve so that speed may be adjusted as needed by certified technician as filter loads
- Two exhaust HEPA filters, 99.99% efficient on particles 0.3 micron in size
- Pressure gauge
- Two utility ports with iris openings
- Tempered safety glass 6.35mm (0.25 in.) thick front sash and sides
- Ergonomic angled and hinged, pivoting sash with wiping seal
Specifications
Balance Enclosure | |
Certifications: ANSI Z9.5, CAN/CSA C22.2, Modified ASHRAE 110, SEFA 1, UL 61010 | |
115 V |
Fluorescent | |
60 Hz |