Learn More
Longwood Elastomers Inc Butyl Rubber Tubing , 1/4 in. ID, 10FT/pack
Supplier: Longwood Elastomers Inc 14452332PK
For use with air and carbon dioxide. Resists oxygen degradation. Heavy wall. Package contains approximately 10 ft. (3m). 3.17 mm thick.