Learn More
Showa Denko America Inc RSpak KC-811
Supplier:  Showa Denko America Inc F6378030
Encompass_Preferred
Columns suitable for the analysis of organic acids, separates compounds by ion exclusion mode and reversed phase mode, highly selective when used with post column method, USP L17 and L22 listed, dimensions: 8.0 mm ID x 300 mm L
Catalog No. 50-161-3268