Learn More
Showa Denko America Inc RSpak JJ-50 2D
Supplier:  Showa Denko America Inc F7008220
Encompass_Preferred
Rugged polyvinyl alcohol packing material is modified with trace amounts of quaternary ammonium groups, and supportsmultimode (reversed phase and anion exchange) analysis, ideal for analysis of complex samples containing neutral and ionic substances, analyses include phenylenediamines and water ion analysis especially for bromate, bromide determination
Catalog No. 50-161-3249