Learn More
Showa Denko America Inc RSpak DS-413
Supplier: Showa Denko America Inc F7001012
Reversed phase analytical column with styrene divinylbenzene base, suitable for reversed phase analysis of highly hydrophilic substances that are not well retained by silica based C18 columns, USP L21 listed, excellent for fatty acid methyl esters, alkylbenzenes and fat soluble vitamins, dimensions are 4.6 mm ID x 150 mm L