Learn More
Hach Company RoVer Rust Remover, pk/12
Supplier: Hach Company 30012
Developed for cleaning rust-fouled ion-exchanged resins. Use to remove rust desposits in lab, shop, laundry or factory applications. Easy-to-use granules pour freely from container. Read Material Safety Data Sheet before using. Case of 12 454 gram bottles.
This item is not returnable.
View return policy