Learn More
Rotronic Instrument Corp Hygroflex3-HF3-HVAC Transmitter
Supplier: Rotronic Instrument Corp HF320WB66XXXX
The HF3 HVAC Transmitter measures relative humidity and temperature and calculates the dew/frost point in HVAC applications focused on relability and repeatability.