Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Rotor S-4x750
Medium capacity swing-bucket rotor with max. speed 4,800 x g (4,700 rpm); includes 4 x 750mL round buckets
$2514.00 - $4763.00
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
13-864-323
|
Eppendorf™
5895120008 |
Rotor S-4x750, incl. 4 × 750 mL round buckets |
Each for $4,763.00
|
|
|||||
13-864-324
|
Eppendorf™
5895121004 |
Rotor S-4x750, without buckets |
Each for $2,514.00
|
|
|||||
Description
- Broad range of adapters enables high versatility for tubes and flasks from 0.2mL to 750mL and plates
- Mixed loading with 2 plate buckets and 2 round tube buckets allows for increased versatility
- Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
- Buckets, caps and adapters are autoclavable
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title