Learn More
Beckman Coulter O-RING., #160., 5.237 ID., 5.443 OD., 0.103 W., 70 DURO., NITRILE
Supplier: Beckman Coulter 878260
O-Ring for Type 45 Ti Rotor Lid, Replacement Rotor Part, 5.443 OD, 0.103W, 70 DURO, Nitrile