Learn More
Hettich Centrifuges ROTANTA 460RF, (24) Plate, Package
Supplier: Hettich Centrifuges 460RFPLATE
Hettich ROTANTA 460 RF PLATE PACKAGE, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters , Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years