Learn More
Electron Microscopy Sciences Rotamix™ RKVSD with Digital Display
SDP
Supplier:  Electron Microscopy Sciences 6105110
Encompass_Preferred
The Rotamix units are designed with interchangeable 12" long rotary rods accommodating tubes ranging from 1.5 ml to 50 ml, which make the rotary universal. Rotamix is made from steel, coated with a chemically resistant paint and is extremely quiet. Rotamix is suitable for use both in the cold room and the incubator. The speed is variable from 0 to 80 rpm. On the RKVSD, rotation speed is displayed on a large LCD. All units are analog controlled and come without racks. Racks are sold separately.
Can be used for immunoprecipitation, extractions and phase separations, bead mixing and extracting. Blood mixing, bacterial cultures, and embedding infiltration.
Catalog No. 50-996-363