Learn More
World Precision Instrument Rongeur, Side Cutter, 15.5cm
Supplier: World Precision Instrument 503226
- 15.5cm (6.1") Long
- Stainless steel
- Used for Trimmins and Recontouring Aleolar Bone and Gross Tissue Removal