Learn More
Braintree Scientific Rodent Jackets, Rat - Large, 350 to 500 grams
Supplier: Braintree Scientific RJL
Rodent Jackets. Manufactured from breathable, latex-free power net mesh in a saddle-style format. Designed for use with rodent tethers and available in a range of sizes to ensure a secure yet comfortable fit. **Sold for research purposes only and not for human use**