Learn More
Electron Microscopy Sciences Acrylic Rodent Brain Matrix Adult Mouse, Sagital
SDP
Supplier:  Electron Microscopy Sciences 69080S
Encompass_Preferred
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
Type: Sagital
Catalog No. 50-292-64