Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Rocking Shaker
OHAUS Rocking Shakers are designed for use in a range of lab applications in a variety of environmental conditions. Includes a non-slip mat.
Supplier: OHAUS™ 30391994/EMD
Description
- Provide a seesaw-like motion
- Electronic tilt adjustment from 0 to 15° while unit is operating
- Displays for temperature, speed and time
- Timer with audible alarm
Specifications
0 to 15° | |
1 to 50 rpm | |
120 V | |
5 A | |
5.5in. | |
16.8 in. | |
42.5 cm | |
10in. | |
25.4 cm | |
15.5 lb. | |
SHRK04DG | |
non skid rubber mat, 3 wire cord and plug |
±1 rpm | |
110 V | |
25 W | |
50/60 Hz | |
14 cm | |
12.8in. | |
32.4 cm | |
10 in. | |
25.4 cm | |
7 kg | |
Digital | |
1 sec. to 160 hr. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction