Learn More
VAISALA INC Humidity and Temperature Transmitter
Supplier:  VAISALA INC HMD62
Encompass_Preferred
For measuring humidity and temperature. The sturdy and reliable duct mounted Vaisala HUMICAP Humidity Transmitter Series is designed for monitoring relative humidity in demanding HVAC and light industrial applications. The combination of high accuracy, stability and reliable operation makes this product series an ideal choice for demanding applications.
Catalog No. NC1690710