Learn More
Biotix™ Robotic aTIP 70μL Non-Filter Pipette Tips
For use with Agilent™ Velocity 11™ VPrep and Bravo, 384-Head platforms.
Supplier: Biotix™ VB00713SC
Description
- Made from clear polypropylene
- Compatible with 96-and 384-well plates
Specifications
70 μL | |
Sterile | |
Autoclavable |
Non-filtered | |
Clear |