Learn More
Medchemexpress LLC HY-103476 5mg Medchemexpress, Ro15-4513 CAS:91917-65-6 Purity:>98%
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Medchemexpress LLC HY1034765MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Medchemexpress, HY-103476 5mg Ro15-4513 CAS:91917-65-6 Ro15-4513, imidazobenzodiazepinone derivative, is a partial inverse agonist of benzodiazepine receptor (BZR). Ro15-4513 is a potent ethanol antagonist. Ro15-4513 has anti-anxiety effect[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.