Learn More
Uline Boston Round Squeezable Bottles - 8 oz, 32/cs
Supplier: Uline S17042CS
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Dispense precise amounts of adhesives, dyes and paints. Soft, translucent LDPE bottles with Yorker cap. Suitable for dropper applications. FDA compliant. Caps included.