Learn More
Supply Solutions RNaseZAP™, 250mL
Supplier: Supply Solutions SIGMA R2020250ML
RNaseZAP™ has been used as a cleaning agent for removing RNase from glassware, apparatus, countertops and pipettors.
This item is not returnable.
View return policy