Learn More
Research Products International Corp RNase Away, 1 Liter Bottle
Supplier: Research Products International Corp 147001
RNase Away removes RNase and DNA contamination from glassware and plastic quickly and economically. Simply apply RNase away to surface or soak as needed and rinse with water. Non-abrasive formulation does not contain strong acids. Effective for use with electrophoresis equipment, glass plates, and other laboratory equipment.
This item is not returnable.
View return policy