Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ringers Rope Rescue Gloves
Click to view available options
Color:
Black
Red
Size:
Small
Medium
Large
X-Large
2X-Large
Specifications
Color | Black |
Material | Neoprene, Synthetic Leather |
Product Type | Extrication Gloves |
Size | Small |
For Use With (Application) | Rescue, Rappelling, and SWAT Situations |
Glove Cuff Style | Hook and Loop |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction