Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyramex™ Ridgeline™ 4-Point Vented Capstyle Hard Hat
Ridgeline™ 4-Point Vented Capstyle Hard Hat is strong, yet ultra-light for seemingly weightless protection.
$217.00 - $617.00
Specifications
For Use With (Application) | Construction |
---|---|
Material | Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam |
Product Type | Vented 4-Point Ratchet |
Size | 6.5 to 8 |
Weight (Metric) | 358 g |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-155-434
|
Pyramex™
HP44110V |
White |
Case of 16 for $225.00
|
|
|||||
19-155-436
|
Pyramex™
HP44112V |
Gray |
Case of 16 for $225.00
|
|
|||||
19-155-439
|
Pyramex™
HP44116SV |
White Graphite |
Case of 16 for $617.00
|
|
|||||
19-155-441
|
Pyramex™
HP44117V |
Matte Black Graphite |
Case of 16 for $617.00
|
|
|||||
19-155-444
|
Pyramex™
HP44120V |
Red |
Case of 16 for $217.00
|
|
|||||
19-155-446
|
Pyramex™
HP44130V |
Yellow |
Case of 16 for $217.00
|
|
|||||
19-155-448
|
Pyramex™
HP44131V |
Hi-vis Lime |
Case of 16 for $282.00
|
|
|||||
19-155-451
|
Pyramex™
HP44141V |
Hi-vis Orange |
Case of 16 for $279.00
|
|
|||||
19-155-453
|
Pyramex™
HP44160V |
Blue |
Case of 16 for $217.00
|
|
|||||
Description
- ABS material – strong, yet ultra-light for seemingly weightless protection
- Low profile design – offers a low center of gravity for better balance
- Rear padded suspension – provides additional level of comfort on the wearers' neck
- 4-position harness points allows you to move the harness forward, backward, up or down – allowing for the most optimal position for ultimate comfort
- Replaceable sweat band – padded fabric and polyurethane foam for all day wear
- Vented pressure pad keeps the straps in place providing an additional level of comfort
- Easy to grasp knob adjusts the suspension from 6 1/2 to 8
Specifications
Construction | |
Vented 4-Point Ratchet | |
358 g | |
Short Brim (Cap Style) |
Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam | |
6.5 to 8 | |
Vented |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title