Learn More
MOLDPRO INC Ribbed Watch Glass for 50 mL Digestion Tube, Clear, 1000/pkg, PP Homopolymer
Supplier: MOLDPRO INC MP121
MP-121 Ribbed Watch Glass is designed to fit all 50 mL digestion vessels. Features large center stem for easy handling. • Made from virgin polypropylene • 1000 pieces/case • Direct replacement for Environmental Express SC505 and SCP Science 010-500-081