Learn More
AG Scientific Inc Ribavirin, 250 MG
Supplier:  AG Scientific Inc R2635250MG
Encompass_Preferred
Synthetic nucleoside analogue structurally related to guanine. Inhibits the replication of a wide range of DNA and RNA viruses.CAS Number: 36791-04-5Molecular Weight: 244.2Chemical Formula: C8H12N4O5Solubility: Soluble in waterStorage Temperature: 0-5CResearch or further manufacturing use only, not for food or drug use."
Catalog No. 50-223-6864