Learn More
Research Products International Corp Ribavirin, 1 G
Supplier: Research Products International Corp R640201.0
Synthetic nucleoside analogue structurally related to guanine. Inhibits the replication of a wide range of DNA and RNA viruses.
WARNING! This product can expose you to Ribavirin, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.