Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ RHINOTUFF™ Puncture Resistant Insoles
Impacto™ RHINOTUFF Puncture Resistant Insoles with stainless steel are designed to fit into most types of work boots and shoes.
Supplier: Impacto™ RHINOTUFFE
Description
Impacto™ Rhinotuff puncture resistant Insoles
- Flexible stainless steel plate to protect from accidental puncture wounds from sharp objects such as pieces of chards, metal and nails
- Molded design with heal cup and arch support for all day support and comfort
- Fabric cover offers protection from moisture
- Used in many industries including construction, manufacturing, fire fighting, forestry, landscaping and mining
Specifications
Red | |
35.56 cm | |
Men: 13 to 14 | |
Rhinotuff Insoles |
14 in. | |
Stainless Steel | |
Unisex |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction