Learn More
iQ Biosciences Rhesus Monkey Splenocytes, 50M cells/vial
Supplier:  iQ Biosciences IQBMNSP203
Encompass_Preferred
High-quality, high-viability (typically > 70%) rhesus monkey (or macaque) splenocytes sourced responsibly from a vetted third-party facility. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor
Catalog No. NC2855591