Learn More
Eisco™ Rheostats
SCI_ED
Wide range of rheostats offering excellent value
Supplier:  Eisco™ PH1150J
 View more versions of this product
Catalog No. S96308
Add to Cart
Description
Description
  • Vitreous enamelled steel tube wound with heavily oxidized resistance wire
  • Supported on robust diecast end stands
  • Heavy-duty siding contact is multileaf phosphor bronze and nickel-plated for corrosion resistance
  • Terminal posts are fitted, facilitating external connection to conventional 0.16 in. (4mm) plug
Specifications
Specifications
Rheostat
36 cm
1.7 in.
14.2 x 4.3 x 4.3 in. (36 x 11 x 11 cm)
11 cm
2.38 lb.
4.3 in.
14.2 in.
1.2 A
4.31 cm
4.3 in.
280 Ω
1.08 kg
11 cm
Product Suggestions
Product Suggestions