Learn More
Eisco™ Rheostats
SCI_ED
Wide range of rheostats offering excellent value
Supplier:  Eisco™ PH1150K
 View more versions of this product
Catalog No. S96305
Add to Cart
Description
Description
  • Vitreous enamelled steel tube wound with heavily oxidized resistance wire
  • Supported on robust diecast end stands
  • Heavy-duty siding contact is multileaf phosphor bronze and nickel-plated for corrosion resistance
  • Terminal posts are fitted, facilitating external connection to conventional 0.16 in. (4mm) plug
Specifications
Specifications
Rheostat
30 cm
1.7 in.
37 Ω
11.8 in.
3.2 A
4.31 cm
Product Suggestions
Product Suggestions