Learn More
Medchemexpress LLC HY-111498A 10mg Medchemexpress, RGX-104 (free Acid) CAS:610318-54-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY111498A10MG
Encompass_Preferred
Medchemexpress, HY-111498A 10mg RGX-104 (free Acid) CAS:610318-54-2 RGX-104 free Acid is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8889