Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RFP Tag (aa 22-36) Synthetic Peptide
Synthetic Peptide
Supplier: Invitrogen™ PEP260
Description
RFP tag (residues 22-36) Synthetic Peptide, PEP-260, from Invitrogen.
PEP-260 is a 15 amino acid synthetic peptide whose sequence is derived from the RFP tag protein. The sequence of this peptide is (amino to carboxy terminus): V (22)-N-G-H-E-F-E-I-E-G-E-G-E-G-R (36)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, RFP Epitope Tag, catalog # PA1-986. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Non-species Specific | |
RFP Tag (aa 22-36) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
No preservative | |
50 μg | |
V(22) - N - G - H - E - F - E - I - E - G - E - G - E - G - R (36) |
purified | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Yes | |
Peptide | |
RUO - research use only |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction