Learn More
Kamiya Biomedical RF (Ver.2) Reagent, 20mL+20mL/PK RX
Supplier: Kamiya Biomedical KAI230
K-ASSAY RF (Ver.2) Assay Reagent. R1: 1 x 20mL (liquid), R2: 1 x 20mL (liquid). Assay Range: 6.65 - 600 IU/mL. The K-ASSAY RF (Ver.2) assay is intended For the quantitative determination of human IgG rheumatoid factor antibodies in patient serum or plasma (citric acid, EDTA, or lithium heparin) based on immunoturbidimetric assay. The presence of IgG RF antibodies, in conjunction with clinical findings and other laboratory tests, is an aid in the diagnosis of rheumatoid arthritis (RA). FOR IN VITRO DIAGNOSTIC USE. Required Calibrator: K-ASSAY RF Calibrator (Ver.2) (Cat. No. KAI-231C). Recommended Controls: K-ASSAY ARC Control (Cat. No. K55C-2M or K55C-4M). This assay is intended for use on most popular chemistry analyzers that allow open channel reagents. Please contact KAMIYA BIOMEDICAL COMPANY to check parameter availability for your analyzer.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.