Learn More
Kamiya Biomedical RF Calibrator (Ver.2), 6x1mL/PK RX
Supplier: Kamiya Biomedical KAI231C
K-ASSAY RF Calibrator (Ver.2). 6 x 1 mL (liquid). For 6 point calibration. The K-ASSAY RF Calibrator (Ver.2) is intended to be used for the calibration of the K-ASSAY RF (Ver.2) immunoturbidimetric assay (Cat. No. KAI-230). FOR IN VITRO DIAGNOSTIC USE.
This item is not returnable.
View return policy