Learn More
LI-COR Revert™ 700 Total Protein Stain for Western Blot Normalization (100 mL)
Supplier: LI-COR 92611011
Make Western blot normalization more accurate and reliable with Revert 700 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 700 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment.Revert 700 Total Protein Stain is a robust and replicable solution for Western blot normalization. The stain can be detected in the 700 nm channel of Odyssey Imaging Systems (or competitor systems) for quantification and normalization analyses. The protocol is quick and can be completed within 15 minutes.The 100 mL size is sufficient for 20 mini Western blots (5 mL/blot), and the 250 mL size is sufficient for 50 mini Western blots.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.