Learn More
Electron Microscopy Sciences Universal Mount Holder
SDP
Supplier:  Electron Microscopy Sciences 76500
Encompass_Preferred
Box with insert and lid; holds 12 of 10 and 15mm diameter mounts or 12 of 3.2mm (1/8") pin mounts.
Catalog No. 50-295-65
Explore available promotions