Learn More
Worthington Biochemical Corporation REVERSE TRANSCRIPTASE 200UN
Supplier: Worthington Biochemical Corporation LS05003
Chromatographically purified dimeric form with M.W. of 66 kDa and 51 kDa. A solution in 10 mM potassium phosphate, pH 7.4, 1 mM DTT and 20% glycerol. 9068-38-6