Learn More
NEOGEN Reveal 2.0 for Listeria
Supplier: NEOGEN 700002796
The Reveal 2.0 for Listeria test provides for the rapid recovery of Listeria in food and environmental samples allowing next day detection and presumptive identification of Listeria.Approvals: AOAC-RI 041101Limit of Detection: 1 cfu/analytical unitTesting Time: 20 MinutesTime to Result: 27-30 HoursFor more information visit https://www.neogen.com/solutions/microbiology/reveal-2-listeria/
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.