Learn More
Research Products International Corp Reusabl Silica Gel Desic Bags
Supplier: Research Products International Corp 420108A
Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators. Durable microfiber bag securely contains beads eliminating the risk of leakage and is unaffected by moisture. Indicator dot changes from blue when dry to pink when moist to easily identify when recharging is needed. Saturated gel can be regenerated by microwaving bag for 6 minutes or Regenerate at 150C (blue color changes to pink_ May be regenerated repeatedly without degradation. Supplied individually sealed in moisture-proof pouch.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.